Learn More
CD45.1 Monoclonal Antibody (A20), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2730.00 NOK
Specifications
Antigen | CD45.1 |
---|---|
Clone | A20 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15590437
|
Affymetrix eBioscience
61-0453-82 |
100 μg |
2730.00 NOK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The A20 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.1-expressing mouse strains. The strains that express CD45.1 include SJL/J, DA, STS/A and RIII. CD45.1 is expressed by all leukocytes in these strains. Applications Reported: This A20 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A20 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD45.1 Monoclonal antibody specifically detects CD45.1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD45.1 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P06800 | |
Ptprc | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Ptprc |
A20 | |
Flow Cytometry | |
PE-eFluor 610 | |
Mouse | |
Mouse | |
19264 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.