Description
The CD44 antibody from Proteintech is a mouse monoclonal antibody generated with purified t cells from human lymph nodes of human CD44. This antibody recognizes human antigen. The CD44 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
Specifications
| Antigen | CD44 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | F10-44-2 |
| Concentration | 10 μL/Test |
| Conjugate | PE |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 960 |
| Show More |