missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (740017) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6577JF669
4437.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CD44 Monoclonal antibody specifically detects CD44 in Rat samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CD44 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Mouse myeloma cell line NS0-derived recombinant rat CD44, Gln22-Thr223 (predicted), Accession # P26051 | |
| RUO | |
| Primary | |
| Rat | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 740017 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 960 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction