missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD42b/GPIb alpha Antibody (SR1163), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21781-50ul
This item is not returnable.
View return policy
Description
CD42b/GPIb alpha Monoclonal antibody specifically detects CD42b/GPIb alpha in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CD42b/GPIb alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1163 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
| Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
| A synthesized peptide derived from human CD42b/GPIb alpha (Uniprot #: P07359) | |
| 50 μg | |
| Immunology | |
| 2811 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction