Learn More
CD40 Monoclonal Antibody (OTI4D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812679S
Encompass_Preferred
 View more versions of this product
Product Code. 17597254
  • 2832.96 NOK / 30µL
Description
CD40 Monoclonal antibody specifically detects CD40 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CD40
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P25942
Mouse
Protein A/G
RUO
958
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry
OTI4D12
Unconjugated
CD40
AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5
Human recombinant protein fragment corresponding to amino acids 21-193+Mouse Fc of human CD40 (NP_001241) produced in 293E cell.
30 μL
Primary
Human
Antibody
IgG2b
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.