Learn More
Invitrogen™ CD314 (NKG2D) Monoclonal Antibody (CX5), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
33410.00 NOK - 150400.00 NOK
Specifications
| Antigen | CD314 (NKG2D) |
|---|---|
| Clone | CX5 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17151332
|
Invitrogen™
16-5882-38 |
5 mg |
33410.00 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17131332
|
Invitrogen™
16-5882-025 |
25 mg |
100600.00 NOK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17141332
|
Affymetrix eBioscience
16-5882-050 |
50 mg | |||||||
Description
The CX5 MAb reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1alpha, -beta, -gamma, -delta, -ε and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1^+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.Specifications
| CD314 (NKG2D) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| CX5 | |
| Flow Cytometry, Functional Assay, Neutralization | |
| Functional Grade | |
| Rat | |
| Mouse | |
| O54709 | |
| 27007 | |
| Primary | |
| 4°C | |
| Klrk1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.