Learn More
CD31 Rat anti-Mouse, Unconjugated, Clone: 390, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65058-1-IG-500UG
Description
Platelet endothelial cell adhesion molecule-1 (PECAM-1, CD31) is a member of the immunoglobulin gene superfamily of cell adhesion molecules. CD31 is a transmembrane glycoprotein that is highly expressed on the surface of the endothelium, making up a large portion of its intracellular junctions. PECAM-1 is also present on the surface of hematopoietic cells and immune cells including platelets, monocytes, neutrophils, natural killer cells, megakaryocytes and some types of T-cell (PMID: 9011572). As well as its role in cell-cell adhesion, PECAM-1 functions as a signaling receptor, and is involved in important physiological events such as nitric oxide production, regulation of T-cell immunity and tolerance, leukocyte transendothelial migration and inflammation and angiogenesis (PMID: 21183735; 20978210; 17872453; 20634489).
Specifications
| CD31 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Primary | |
| Mouse | |
| IgG2a |
| Immunofluorescence, Flow Cytometry | |
| 390 | |
| Unconjugated | |
| 500 μg | |
| 18613 | |
| Liquid |