missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 epsilon Antibody (145-2C11) [Janelia Fluor™ 669], Novus Biologicals™
Hamster Monoclonal Antibody
Brand: Novus Biologicals FAB484JF669
4772.75 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Cell depletion,T Cell Stimulation,CyTOF-reported
Specifications
| CD3 epsilon | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| H-2Kb-specific mouse cytotoxic T-lymphocyte | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Depletion Assay, T-Cell Activation, CyTOF | |
| 145-2C11 | |
| 50mM Sodium Borate | |
| Hamster | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 916 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction