missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD3 delta Polyclonal antibody specifically detects CD3 delta in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CD3 delta |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse CD3 delta. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?