Learn More
Invitrogen™ CD3/CD19 Antibody Cocktail, FITC, PE, eBioscience™
Description
Description: This product consists of two fluorochrome-conjugated monoclonal antibodies to distinguish human peripheral CD3^+ T cells and CD19^+ B cells. The UCHT1 monoclonal antibody (mouse IgG1, kappa) reacts with human CD3e, a 20 kDa subunit of the TCR complex.Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The HIB19 monoclonal antibody (mouse IgG1, kappa) reacts with human CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells.Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. Applications Reported: The product is formulated for use in flow cytometric analysis. Applications Tested: This product has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 20 μL per test (containing 1 μg of Anti-Human CD3 (UCHT1) FITC and 0.25 μg of Anti-Human CD19 (HIB19) PE). A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number...
Specifications
Specifications
| Antigen | CD3/CD19 |
| Applications | Flow Cytometry |
| Classification | Cocktail |
| Clone | UCHT1, HIB19 |
| Concentration | 20 μL/Test |
| Conjugate | FITC/PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD3G |
| Gene Accession No. | P04234, P07766, P09693, P15391, P20963 |
| Gene Alias | 4930549J05Rik; A430104F18Rik; AI504783; AW495831; AW552088; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; CD19; CD19 antigen; CD19 molecule; Cd247; CD247 antigen; CD247 antigen, zeta subunit; CD247 molecule; CD3; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; Cd3d; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; CD3E; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3epsilon; CD3-epsilon; Cd3-eta; CD3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; Cd3-zeta; CD3zeta chain; CD3-zeta/eta; Ctg3; Ctg-3; CVID3; Differentiation antigen CD19; FLJ18683; IMD17; IMD18; IMD19; IMD25; OKT3, delta chain; T3D; T3E; T3G; T3Z; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor T3 delta chain; T-cell receptor T3 eta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen Leu-12; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; TCR zeta chain; TCRE; Tcrk; Tcrz |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.