missing translation for 'onlineSavingsMsg'
Learn More

CD28 Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA813688

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17558394

  • 4065.15 NOK / 100µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD28
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P10747
Mouse
Protein A/G
RUO
940
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry
OTI1A2
Unconjugated
Cd28
antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44
Full length human recombinant protein of human CD28 (NP_006130) produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers