missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Antibody (37407) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB342JF525
4853.50 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Western Blot,Agonist Activity,Immunocytochemistry
Specifications
| CD28 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD28, Asn19-Pro152, Accession # P10747 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 940 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Agonist Activity, Immunocytochemistry | |
| 37407 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction