Learn More
CD279 (PD-1) Mouse anti-Human, PE, Clone: J110, Proteintech
Product Code. 16821716 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16821716 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16821716 Supplier Proteintech Supplier No. PE65119100TESTS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mouse Monoclonal Antibody
The CD279 (PD-1) antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279 (PD-1). This antibody recognizes human antigen. The CD279 (PD-1) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
Antigen CD279 (PD-1)
Applications Flow Cytometry
Classification Monoclonal
Clone J110
Concentration 5 μL (1 μg)/Test
Conjugate PE
Host Species Mouse
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human
Isotype IgG1