Learn More
CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5983-38
Description
The MIH1 monoclonal antibody reacts with human B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH1 is reported to be a blocking antibody. Applications Reported: The MIH1 antibody has been reported for use in flow cytometric analysis. It has also been reported in blocking in in vitro functional assays. Applications Tested: The MIH1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD274 (PD-L1, B7-H1) Monoclonal antibody specifically detects CD274 (PD-L1, B7-H1) in Human samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
Specifications
| CD274 (PD-L1, B7-H1) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q9NZQ7 | |
| Mouse | |
| 5 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Functional Assay, Neutralization | |
| MIH1 | |
| Functional Grade | |
| Cd274 | |
| A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; CD274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; PDCD1L1; PDCD1LG1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
| Affinity chromatography | |
| RUO | |
| 29126 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.