missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Monoclonal Antibody (OTI8E5), TrueMAB™, OriGene
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
| Antigen | CD27 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI8E5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | CD27 |
| Gene Accession No. | P26842 |
| Gene Alias | CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?