missing translation for 'onlineSavingsMsg'
Learn More

CD27 Monoclonal Antibody (OTI5C10), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA812553S

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17537044

  • 4065.15 NOK / 30µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

CD27
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P26842
Mouse
Protein A/G
RUO
939
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Western Blot
OTI5C10
Unconjugated
CD27
CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7
Full length human recombinant protein of human CD27 (NP_001233) produced in HEK293T cell.
30 μL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers