Learn More
CD24 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18330-1-AP-150UL
This item is not returnable.
View return policy
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.Specifications
| CD24 | |
| Polyclonal | |
| Unconjugated | |
| CD24 | |
| CD24, CD24 molecule, CD24A, Signal transducer CD24 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100133941, 12484 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24807, P25063 | |
| CD24 | |
| CD24 Fusion Protein Ag11679 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |