missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 Mouse anti-Human, Clone: 7A2F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66103-1-IG-20UL
This item is not returnable.
View return policy
Description
CD22 (BL-CAM) is a type 1 integral membrane glycoprotein with molecular weight of 130 to 140 kDa. CD22 is expressed in both the cytoplasm and cell membrane of B-lymphocytes. CD22 antigen appears early in B-cell lymphocyte differentiation at approximately the same stage as the CD19 antigen. Unlike other B-cell markers, CD22 membrane expression is limited to the late differentiation stages comprised between mature B cells (CD22+) and plasma cells (CD22-), and may thus prove useful in phenotyping mature leukemias. CD22 is also strongly expressed in hairy cell leukemia. CD22 preferentially binds to alpha2,6-linked sialic acid. The sialic acid recognition site can be masked by cis interactions with sialic acids on the same cell surface. Upon ligand induced tyrosine phosphorylation in the immune response seems to be involved in regulation of B cell antigen receptor signaling. CD22 plays a role in positive regulation through interaction with Src family tyrosine kinases and may also act as an inhibitory receptor by recruiting cytoplasmic phosphatases via their SH2 domains that block signal transduction through dephosphorylation of signaling molecules. CD22 is also strongly expressed in hairy cell leukemia. CD22 is also positive in diffuse large B-cell lymphoma and nodular lymphocyte predomice Hodgkin′s lymphoma, but negative in classical Hodgkin′s lymphoma.Specifications
| CD22 | |
| Monoclonal | |
| 2.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20273 | |
| CD22 | |
| CD22 Fusion Protein Ag17986 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 7A2F1 | |
| Unconjugated | |
| CD22 | |
| B cell receptor CD22, BL CAM, CD22, CD22 molecule, SIGLEC 2, SIGLEC2, T cell surface antigen Leu 14 | |
| Mouse | |
| Protein A | |
| RUO | |
| 933 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction