Learn More
CD21 Monoclonal Antibody (OTI3E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2832.96 NOK - 6331.68 NOK
Specifications
| Antigen | CD21 |
|---|---|
| Clone | OTI3E1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17548974
|
Origene Technologies
TA814205S |
30 μL |
2832.96 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17528974
|
Origene Technologies
TA814205 |
100 μL |
6331.68 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD21 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
| Human recombinant protein fragment corresponding to amino acids 208-587 of human CD21 (NP_001868) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| CR2 |
| OTI3E1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P20023 | |
| 1380 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.