Learn More
CD205 Monoclonal Antibody (205yekta), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1132.00 NOK - 2715.00 NOK
Specifications
Antigen | CD205 |
---|---|
Clone | 205yekta |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15372010
|
Affymetrix eBioscience
53-2051-82 |
100 μg |
2715.00 NOK
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15362010
|
Affymetrix eBioscience
53-2051-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells. Applications Reported: This 205yekta antibody has been reported for use in flow cytometric analysis. Applications Tested: This 205yekta antibody has been tested by flwo cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD205 Monoclonal antibody specifically detects CD205 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD205 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q60767 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
205yekta | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
17076 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.