missing translation for 'onlineSavingsMsg'
Learn More

CD200 Monoclonal Antibody (OX104), Biotin, eBioscience™, Invitrogen™

Product Code. p-7192840
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Product Code. Quantity unitSize
17190392 25 μg 25µg
17100402 100 μg 100µg
2 options
This item is not returnable. View return policy

Product Code. 17190392

Brand: Invitrogen 13920080

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: The monoclonal antibody OX104 recognizes human CD200 also known as OX2. CD200 is a member of the Ig superfamily with 2 Ig domains, a transmembrane and cytoplasmic domain. CD200 is expressed on resting and activated B cells, a subset of resting and activated T cells, keratinocytes, peripheral and central nerve cells, follicular dendritic cells and ovarian cells. The interaction with CD200R results in macrophage activation (IL-6 production), inhibition of mast cell degranulation along with reduced TNFalpha and IL-13 secretion and overall attenuation of the activation status of lymphocytes. A role has also been suggested in maternal tolerance as expression of CD200 is also present on the trophoblast. Applications Reported: This OX104 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX104 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

CD200 is a type-1 membrane glycoprotein that contains two immunoglobulin domains, and belongs to the immunoglobulin superfamily. CD200 is a broadly distributed cell surface glycoprotein that interacts with a receptor on myeloid cells (CD200R) and is implicated in locally preventing macrophage activation. Distant, but recognizable, homologues of CD200 have been identified in many herpesviruses and poxviruses. Studies of the related genes in mouse and rat suggest that CD200 may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms of CD200 have been found. Diseases associated with CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200
Applications Flow Cytometry
Classification Monoclonal
Clone OX104
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cd200
Gene Accession No. P41217
Gene Alias antigen identified by monoclonal antibody MRC OX-2; BOS_763; CD200; CD200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein
Gene Symbols Cd200
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4345
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.