Learn More
CD19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27949-1-AP-20UL
 View more versions of this product
Product Code. 16805925
  • 1710.33 NOK / 20µL
Description
CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lamda light chains. It appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and maligt B cell proliferation. It is expressed in all B cell precursor leukemias.
Specifications
Specifications
CD19
Polyclonal
Unconjugated
CD19
B lymphocyte antigen CD19, B4, CD19, CD19 molecule, Differentiation antigen CD19, T cell surface antigen Leu 12
Rabbit
Antigen Affinity Chromatography
RUO
930
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P15391
CD19
CD19 Fusion Protein Ag27656