Learn More
CD180 (RP105) Monoclonal Antibody (MHR73-11), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 16-1809-050
 View more versions of this product
Product Code. 17101182
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The MHR73-11 MAb reacts with human CD180 (RP105). This 105 kDa type I transmembrane molecule is a member of the TLR family of proteins characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. RP105 physically associates with another molecule called MD-1 and is expressed on B, monocytes/macrophages, and dendritic cells. Histological studies show that RP105 is expressed mainly on mature B cells in mantle zones, while germinal center cells are either dull or negative. The RP105/MD-1 complex in concert with TLR4 mediates B cell recognition and signaling of LPS. MHR73-11 activates B cells, leading to increases in cell size, expression of the costimulatory molecule CD80, and DNA synthesis. Moreover, ligation of RP105 protects B cells from irradiation- or dexamethasone-induced apoptosis. Thus, RP105 is a signal transduction molecule and plays a role in regulation of B cell growth and death. A significant proportion of circulating B cells in SLE patients is RP105 negative. Loss of RP105 is associated with B cell activation and increased disease activity in SLE patients.
CD180 (RP105) Monoclonal antibody specifically detects CD180 (RP105) in Human samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD180 (RP105)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
Q99467
Mouse
50 mg
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Functional Assay
MHR73-11
Functional Grade
Cd180
Cd180; CD180 antigen; CD180 molecule; F630107B15; LY64; Ly78; Ly-78; lymphocyte antigen 64; Lymphocyte antigen 78; lymphocyte antigen-64, radioprotective, 105kDa; MGC126233; MGC126234; Radioprotective 105 kDa protein; RP105
Affinity chromatography
RUO
4064
4° C