Learn More
CD16 Monoclonal Antibody (3G8), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
3585.00 NOK
Specifications
Antigen | CD16 |
---|---|
Clone | 3G8 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15589086
|
Affymetrix eBioscience
46-0166-42 |
100 Tests |
3585.00 NOK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This 3G8 monoclonal antibody reacts with human and non-human primate CD16, which is also known as the low-affinity Fc gamma RIII. CD16 exists as two distinct isoforms, Fc gamma RIIIA and Fc gamma RIIIB. In humans, Fc gamma RIIIA is expressed as a polypeptide-anchored form on monocytes, macrophages, and lymphocytes such as NK cells. T and B cells do not express this Fc receptor. Fc gamma RIIIB is also detected on neutrophils as a GPI-anchored form. Expression of CD16 on lymphocytes and monocytes is similar in non-human primates. However, while CD16 is not found on neutrophils in macaques and baboons, this receptor is detected on these cells in sooty mangabeys. Binding of IgG leads to activation of signal transduction pathways, resulting in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, cytokine release, and antigen presentation. This monoclonal antibody has been reported to have several functional activities, including inhibition of cytotoxic ability, activation of cell signaling, and NK cell depletion in vivo. Moreover, the 3G8 antibody clone has been demonstrated to work on capuchin monkey, chimpanzee, common marmoset, cynomologous monkey, hamadyras baboon, olive baboon, pigtailed macaque, rhesus, and squirrel monkey. Based on cross-blocking studies 3G8 recognizes the same epitope as CB16. However, 3G8 and B73.1 antibody clones bind distinct epitopes.
CD16 encodes a receptor that recognizes the Fc portion of immunoglobulin G and is involved in the clearance of immune complexes from the circulation, as well as other functions such as cellular mediated cytotoxicity and enhancement of virus infections. This gene, FCGR3A, shares a high degree of similarity with another nearby gene, FCGR3B, located on chromosome 1. The receptor encoded by this gene is expressed on natural killer (NK) cells as an integral membrane glycoprotein anchored through a transmembrane peptide, while FCGR3B is expressed on polymorphonuclear neutrophils (PMN) where the receptor is anchored through a phosphatidylinositol (PI) linkage. Mutations in this gene have been associated with immunodeficiency 20 and have been linked to susceptibility to recurrent viral infections, susceptibility to systemic lupus erythematosus, and alloimmune neonatal neutropenia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. Diseases associated with FCGR3A include Immunodeficiency 20 and Herpes Zoster.Specifications
CD16 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 | |
Affinity chromatography | |
Antibody |
3G8 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
FCGR3A |
For Research Use Only.