missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD151 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10418-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It is involved in cellular processes including cell adhesion and may regulate integrin trafficking and/or function. This protein enhances cell motility, invasion and metastasis of cancer cells. Multiple alternatively spliced transcript variants that encode the same protein have been described for this gene.Specifications
| CD151 | |
| Polyclonal | |
| Unconjugated | |
| CD151 | |
| CD151, CD151 antigen, GP27, Membrane glycoprotein SFA 1, MER2, PETA 3, RAPH, SFA1, Tetraspanin 24, Tspan 24, TSPAN24 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12476, 64315, 977 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35566, P48509, Q9QZA6 | |
| Cd151 | |
| CD151 Fusion Protein Ag0341 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction