missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD151 Antibody (455807) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4609JF585
This item is not returnable.
View return policy
Description
CD151 Monoclonal antibody specifically detects CD151 in Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| CD151 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
| HEK293 human embryonic kidney cell line transfected with mouse CD151, Met1-Tyr253, Accession # BAA22447 | |
| RUO | |
| 977 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 455807 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction