missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD147 Monoclonal Antibody (OX114), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517931
Additional Details : Weight : 0.01000kg
Description
MA5-17931 targets CD147/BSG in FACS applications and shows reactivity with Mouse samples. The MA5-17931 immunogen is recombinant md0-2rCD4d3+4.
CD147 Monoclonal antibody specifically detects CD147 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD147 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P18572 | |
BSG | |
Recombinant md0-2rCD4d3+4 | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX114 | |
PE | |
BSG | |
5A11; 5A11 antigen; 5A11/Basigin; 5A11/Basigin-2; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); blood-brain barrier HT7 antigen; BSG; CD147; CD147 antigen; CE9; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; glycoprotein CE9; gp 42; gp42/basigin/OX-47/HT7; HT7; HT-7; HT7 antigen; immunoglobulin gene superfamily member; leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; Ox47R; RPE7; TCSF; tumor cell-derived collagenase stimulatory factor; Unknown (protein for MGC:128392); unnamed protein product; UNQ6505/PRO21383 | |
Rat | |
Protein G | |
RUO | |
12215 | |
4° C, do not freeze | |
Liquid |