Learn More
CD133-1,2,4,7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19945-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The CD133-1, 2, 4, 7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human CD133-1, 2, 4, 7. This antibody recognizes human, mouse antigen. The CD133-1, 2, 4, 7 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
CD133-1,2,4,7 | |
Polyclonal | |
Unconjugated | |
150 μL | |
8842 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
167 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |