Learn More
Invitrogen™ CD120a (TNF Receptor I) Monoclonal Antibody (55R-170), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
33200.00 NOK - 166100.00 NOK
Specifications
| Antigen | CD120a (TNF Receptor I) |
|---|---|
| Clone | 55R-170 |
| Concentration | 1 mg/mL |
| Applications | Functional Assay, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17161122
|
Invitrogen™
16-1202-38 |
5 mg |
33200.00 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17141122
|
Affymetrix eBioscience
16-1202-025 |
25 mg | |||||||
|
17151122
|
Invitrogen™
16-1202-050 |
50 mg |
166100.00 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 55R-170 MAb reacts with mouse Tumor Necrosis Factor Receptor I (TNFRI, TNFR-p55, TNFRSF1A, CD120a). TNFRI is expressed in a variety of cell types. The soluble TNF receptors are truncated forms of cell surface receptors with neutralizing activity on both TNF-alpha and TNF-beta. It has been suggested that the proinflammatory and immunosuppressive properties of TNF segregate at the level of its receptors and that the pool of TNF bound to soluble receptors could represent a reservoir to control the TNF release. Applications Reported: The 55R-170 antibody has been reported for use in ELISA, neutralization of mouse TNFR I bioactivity and flow cytometric analysis. Applications Tested: The Functional Grade Purified 55R-170 antibody has been tested by LAL assay to verify low endotoxin levels and in bioassay for neutralization of mouse TNFR I bioactivity. The 55R-170 antibody at 25 μg/mL has been found to inhibit by 50% the biological effects of 1 ng/mL mouse TNF alpha in an assay of TNF alpha induced cytotoxicity of L929 cells. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the Best Protocols section. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
TNFR1 (Tumor necrosis factor receptor 1) belongs to the tumor necrosis factor superfamily, and is one of the major TNF-alpha receptors. TNFR1 plays an important role in mediating, in cytokine mediated signaling, positive regulation of the NF-Kb pathway, positive regulation of angiogenesis, and negative regulation of gene expression. The extracellular domain of TNFR1 is also released into the circulatory system as soluble TNFR1 (sTNFR1). In humans, the TNFR1 gene is located on chromosome 12. Anti-apoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with TNFR1, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of TNFR1 were found to be associated with the autosomal dominant periodic fever syndrome, and the impaired receptor clearance is thought to be a mechanism of the disease.Specifications
| CD120a (TNF Receptor I) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 21937 | |
| Primary | |
| 4°C | |
| Tnfrsf1a |
| 55R-170 | |
| Functional Assay, Neutralization | |
| Functional Grade | |
| Armenian Hamster | |
| Mouse | |
| P25118 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.