missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c/Integrin alpha X Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17342-1-AP-150UL
This item is not returnable.
View return policy
Description
CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.Specifications
| CD11c/Integrin alpha X | |
| Polyclonal | |
| Unconjugated | |
| ITGAX | |
| CD11C, Integrin alpha X, ITGAX, Leu M5, SLEB6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3687 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20702 | |
| ITGAX | |
| CD11c/Integrin alpha X Fusion Protein Ag11380 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction