Learn More
CD11b Monoclonal Antibody (ICRF44), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2965.00 NOK
Specifications
Antigen | CD11b |
---|---|
Clone | ICRF44 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15589076
|
Affymetrix eBioscience
46-0118-42 |
100 Tests |
2965.00 NOK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The ICRF44 monoclonal antibody reacts with human CD11b, a 165 kDa adhesion molecule. CD11b associated with integrin beta2 (CD18) is expressed on the surface of monocytes, granulocytes, activated lymphocytes and a subset of NK cells. CD11b is a receptor for intercellular adhesion molecule family members CD54, CD102 and CD50 as well as for iC3b. These adhesions are crucial in cell-cell and cell-matrix interactions. Applications Reported: This ICRF44 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ICRF44 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD11b Monoclonal antibody specifically detects CD11b in Baboon, Chimpanzee, Cynomolgus Monkey, Human, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
CD11b | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P11215 | |
ITGAM | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ITGAM |
ICRF44 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Rhesus Monkey | |
101866035, 3684, 454069, 714832 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.