missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The CD107a antibody from Proteintech is a mouse monoclonal antibody generated with human adherent peripheral blood cells of human CD107a. This antibody recognizes human antigen. The CD107a antibody has been validated for the following applications: IF, FC analysis. This antibody is fitc.
Specifications
Specifications
| Antigen | CD107a |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | H4A3 |
| Concentration | 10 μL/Test |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 3916 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?