missing translation for 'onlineSavingsMsg'
Learn More

CD100 (Sema4D) Rat anti-Mouse, BUV661, Clone: BMA-12, BD Biosciences™

Product Code. 17201845
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
17201845 50 μg 50µg
1 options
This item is not returnable. View return policy

Product Code. 17201845

Brand: BD Biosciences 750506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD100 (Sema4D) Monoclonal antibody specifically detects CD100 (Sema4D) in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD100 (Sema4D)
Applications Flow Cytometry
Classification Monoclonal
Clone BMA-12
Conjugate Brilliant Ultraviolet 661
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Semaphorin-4D; SEM4D; Semaphorin-C-like 2; Semacl2; Semaj; M-Sema G; coll-4
Host Species Rat
Immunogen Mouse CD100 Recombinant Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody.
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.