Learn More
CD 3254, TRC
Click to view available options
Quantity:
1 mg
Specifications
Specifications
| Chemical Name or Material | CD 3254 |
| CAS | 196961-43-0 |
| Molecular Formula | C24H28O3 |
| Quantity | 1 mg |
| Synonym | 3-[4-Hydroxy-3-(5,6,7,8-tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)phenyl]-2-propenoic acid |
| InChI Formula | InChI=1S/C24H28O3/c1-15-12-19-20(24(4,5)11-10-23(19,2)3)14-17(15)18-13-16(6-8-21(18)25)7-9-22(26)27/h6-9,12-14,25H,10-11H2,1-5H3,(H,26,27)/b9-7+ |
| SMILES | CC1(C2=C(C(C)(CC1)C)C=C(C(C)=C2)C3=C(C=CC(/C=C/C(O)=O)=C3)O)C |
| IUPAC Name | (E)-3-[4-hydroxy-3-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)phenyl]prop-2-enoic acid |
| Molecular Weight (g/mol) | 364.48 |