missing translation for 'onlineSavingsMsg'
Learn More

CCT6B Mouse anti-Human, Unconjugated, Clone: 2F11A9, Proteintech

Product Code. 17521476 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17521476 20 μL 20µL
17511476 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17521476

Brand: Proteintech 679611IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CCT6B antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CCT6B. This antibody recognizes human, mouse, rat antigen. The CCT6B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.

Specifications

Antigen CCT6B
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 2F11A9
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 10693
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.