Description
The CCT6B antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CCT6B. This antibody recognizes human, mouse, rat antigen. The CCT6B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
| Antigen | CCT6B |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 2F11A9 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 10693 |
| Target Species | Human, Mouse, Rat |
| Show More |