Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J303, Paragon Scientific™
Product Code. 16040010
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 16040010
Brand: Paragon Scientific CL09
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Manufactured in the United Kingdom
  • 2 year shelf life
Specifications
Packaging Glass Amber Bottle
Certificate of Analysis Yes
Physical Form Liquid
Quantity 500 mL
compliance-icons
Product Identifier
  • CCS Viscosity Standard, CL090 (CL09)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Dec-1-ene, dimers, hydrogenated
Nominal values quoted. Certified Values will vary from batch to batch.