Learn More
CCR5 Mouse anti-Human, Clone: CTC5, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems MAB1802-SP
Encompass
Product Code. 15794554
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.