Learn More
CCL4 (MIP-1 beta) Monoclonal Antibody (FL34Z3L), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1118.00 NOK - 3115.00 NOK
Specifications
Antigen | CCL4 (MIP-1 beta) |
---|---|
Clone | FL34Z3L |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-eFluor 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15589366
|
Affymetrix eBioscience
46-7540-42 |
100 Tests |
3115.00 NOK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15579366
|
Affymetrix eBioscience
46-7540-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CCL4, also known as macrophage inflammatory protein 1-beta (MIP1B), belongs to the intercrine beta (chemokine CC) family. Both CCL4 and the related protein CCL3 participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells. While both CCL4 and CCL3 exert similar effects on monocytes, their effect on lymphocytes differ, with CCL4 selectively attracting CD4+ lymphocytes and CCL3 selectively attracting CD8+ lymphocytes. Additionally, both have been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. The processed form of CCL4 can induce down-modulation of surface expression of the chemokine receptor CCR5, thus inhibiting the CCR5-mediated entry of HIV-1 in T cells.
CCL4 (MIP-1 beta) Monoclonal antibody specifically detects CCL4 (MIP-1 beta) in Human samples. It is validated for Flow Cytometry.Specifications
CCL4 (MIP-1 beta) | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
6351 | |
IgG2a κ | |
Affinity chromatography | |
CCL4 |
FL34Z3L | |
Monoclonal | |
Liquid | |
RUO | |
P13236 | |
CCL4 | |
Primary | |
Antibody |
For Research Use Only.