missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL18/PARC Antibody (64507) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB394JF585
This item is not returnable.
View return policy
Description
CCL18/PARC Monoclonal antibody specifically detects CCL18/PARC in Human samples. It is validated for Western Blot,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| CCL18/PARC | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Alternative macrophage activation-associated CC chemokine 1, AMAC1CC chemokine ligand 18, AMAC-1Small-inducible cytokine A18, chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated), CKb7, DC-CK1CC chemokine PARC, DCCK1Macrophage inflammatory protein 4, Dendritic cell chemokine 1, MIP4, MIP-4C-C motif chemokine 18, PARCPulmonary and activation-regulated chemokine, SCYA18chemokine (C-C), dendritic, small inducible cytokine A18, small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary andactivation-regulated | |
| E. coli-derived recombinant human CCL18/PARC, Ala21-Ala89, Accession # P55774.1 | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer | |
| 6362 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF | |
| 64507 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction