missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL16/HCC-4/LEC Antibody (70218) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB328JF585
This item is not returnable.
View return policy
Description
CCL16/HCC-4/LEC Monoclonal antibody specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
Specifications
| CCL16/HCC-4/LEC | |
| Monoclonal | |
| Janelia Fluor 585 | |
| C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16 | |
| E. coli-derived recombinant human CCL16/HCC-4, Gln24-Gln120, Accession # O15467 | |
| 0.5 mL | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6360 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 70218 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction