missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCKBR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16549-1-AP-150UL
This item is not returnable.
View return policy
Description
CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.Specifications
| CCKBR | |
| Polyclonal | |
| Unconjugated | |
| CCKBR | |
| CCK B, CCK B receptor, CCK BR, CCK2 R, CCK2R, CCKBR, CCKRB, Cholecystokinin 2 receptor, cholecystokinin B receptor, GASR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 887 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32239 | |
| CCKBR | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction