Learn More
CCG-63802, MedChemExpress
Product Code. 16467936
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16467936
Brand: Medchem Express HY70074
MedChemExpress CCG-63802 is a selective, reversible and allosteric RGS4 inhibitor. CCG-63802 specifically binds to RGS4 and blocks the RGS4-Gαo interaction, with an IC50 value of 1.9 μM.
CCG-63802 is a selective, reversible and allosteric RGS4 inhibitor. CCG-63802 specifically binds to RGS4 and blocks the RGS4-Gαo interaction, with an IC50 value of 1.9 μM.
Specifications
Chemical Name or Material CCG-63802
CAS 620112-78-9
Color Orange
Purity Grade Notes Research
Molecular Formula C26H18N4O2S
Quantity 100 mg
Solubility Information DMSO : 1.67 mg/mL (3.71 mM; Need ultrasonic and warming)
SMILES N#C/C(C1=NC2=CC=CC=C2S1)=C\C3=C(N=C4C(C)=CC=CN4C3=O)OC5=CC=CC(C)=C5
Molecular Weight (g/mol) 450.51
Formula Weight 450.51
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.