missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC59 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26387-1-AP-20UL
This item is not returnable.
View return policy
Description
Component of the transcription complexes of the pulmonary surfactant-associated protein-B (SFTPB) and -C (SFTPC). Enhances homeobox protein Nkx-2.1-activated SFTPB and SFTPC promoter activities.Specifications
| CCDC59 | |
| Polyclonal | |
| Unconjugated | |
| CCDC59 | |
| BR22, CCDC59, HSPC128, TAP26, TTF 1 associated protein 26, TTF 1 associated protein BR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29080 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9P031 | |
| CCDC59 | |
| CCDC59 Fusion Protein Ag23852 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction