missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16636-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
CCDC22 is a recently identified coiled-coil domain-containing protein that has been shown to bind copines, which are calcium-dependent, membrane-binding proteins that may function in calcium signaling. In rat, CCDC22 has been observed to localize in multiple regions of the brain, including the prefrontal and somatosensory cortex, dentate gyrus and thalamus, and in the ipsilateral motor neurons of the spinal cord after sciatic nerve transection suggesting that it may play a role in neuronal injury response. The human CCDC22 gene has been identified as a novel candidate gene for syndromic X-linked intellectual disability (XLID).Specifications
CCDC22 | |
Polyclonal | |
Unconjugated | |
CCDC22 | |
CCDC22, CXorf37, JM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28952, 317381, 54638 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60826, P86182, Q9JIG7 | |
Ccdc22 | |
CCDC22 Fusion Protein Ag9980 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |