missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC102A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25764-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifica
CCDC102A | |
Polyclonal | |
Unconjugated | |
CCDC102A | |
CCDC102A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
92922 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96A19 | |
CCDC102A | |
CCDC102A Fusion Protein Ag22753 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |