Learn More
CC-671, MedChemExpress
Product Code. 16496947
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16496947 5 mg 5mg
16484228 10 mg 10mg
16480567 50 mg 50mg
16461109 1 mg 1mg
16435176 100 mg 100mg
5 options
Product Code. 16496947
Brand: Medchem Express HY108709
MedChemExpress CC-671 is a dual TTK protein kinase/CDC2-like kinase (CLK2) inhibitor with IC50s of 0.005 and 0.006 μM for TTK and CLK2, respectively.
CC-671 is a dual TTK protein kinase/CDC2-like kinase (CLK2) inhibitor with IC50s of 0.005 and 0.006 μM for TTK and CLK2, respectively.
Specifications
Chemical Name or Material CC-671
CAS 1618658-88-0
Color Light Green
Purity Grade Notes Research
Molecular Formula C28H28N6O4
Quantity 5 mg
Solubility Information DMSO : ≥ 60 mg/mL (117.06 mM)
SMILES O=C(NC)C1=CC=C(NC2=NC(NC=C3C4=CC(OC(C)=N5)=C5C=C4)=C3C(OC6CCCC6)=N2)C(OC)=C1
Molecular Weight (g/mol) 512.56
Formula Weight 512.56
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease