missing translation for 'onlineSavingsMsg'
Learn More

CBX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11831-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16849163

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is a rabbit polyclonal antibody raised against a full-length human HP1A protein, and can reacts with the 28 kDa HP1A protein.

This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.
Specifications

Specifications

CBX5
Polyclonal
Unconjugated
CBX5
Antigen p25, CBX5, Chromobox protein homolog 5, HP1, HP1 alpha, HP1A
Rabbit
Antigen Affinity Chromatography
RUO
12419, 23468, 300266
-20°C
Liquid
Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P45973, Q61686
CBX5
CBX5 Fusion Protein Ag2403
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers