Learn More
CBX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11831-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against a full-length human HP1A protein, and can reacts with the 28 kDa HP1A protein.
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.Specifications
| CBX5 | |
| Polyclonal | |
| Unconjugated | |
| CBX5 | |
| Antigen p25, CBX5, Chromobox protein homolog 5, HP1, HP1 alpha, HP1A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12419, 23468, 300266 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P45973, Q61686 | |
| CBX5 | |
| CBX5 Fusion Protein Ag2403 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |