Learn More
Abnova™ CBX3 Recombinant Protein
Human CBX3 full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00011335-P01.25ug
Additional Details : Weight : 0.00010kg
Description
At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane. Two transcript variants encoding the same protein but differing in the 5' UTR, have been found for this gene.
- Theoretical MW (kDa): 45.87
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MASNKTTLQKMGKKQNGKSKKVEEAEPEEFVVEKVLDRRVVNGKVEYFLKWKGFTDADNTWEPEENLDCPELIEAFLNSQKAGKEKDGTKRKSLSDSESDDSKSKKKRDAADKPRGFARGLDPERIIGATDSSGELMLLMKWKDSDEADLVLAKEANMKCPQIVIAFYEERLTWHSCPEDEAQ
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH00954 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
45.87 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MASNKTTLQKMGKKQNGKSKKVEEAEPEEFVVEKVLDRRVVNGKVEYFLKWKGFTDADNTWEPEENLDCPELIEAFLNSQKAGKEKDGTKRKSLSDSESDDSKSKKKRDAADKPRGFARGLDPERIIGATDSSGELMLLMKWKDSDEADLVLAKEANMKCPQIVIAFYEERLTWHSCPEDEAQ | |
HECH/HP1-GAMMA/HP1Hs-gamma | |
CBX3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
11335 | |
CBX3 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CBX3 | |
Human | |
Recombinant | |
Solution |